Skip to Content
Merck
All Photos(1)

Key Documents

880135P

Avanti

DSPE-PEG(2000) Carboxylic Acid

Avanti Research - A Croda Brand 880135P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[carboxy(polyethylene glycol)-2000] (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C131H257NNaO55P
CAS Number:
Molecular Weight:
2780.38
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (880135P-10mg)
pkg of 1 × 100 mg (880135P-100mg)
pkg of 1 × 25 mg (880135P-25mg)
pkg of 1 × 50 mg (880135P-50mg)
pkg of 1 × 500 mg (880135P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880135P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(C/C=C\OOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOCCOOCC(O)=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=

General description

Carboxylic acids are readily soluble in polar solvents such as water and alcohols. DSPE-PEG(2000) carboxylic acid consists of a hydrophilic polyethylene glycol (PEG) chain of 2kDa.

Application

DSPE-PEG(2000) carboxylic acid has been used:
  • to covalently conjugate integrin α-2 (ITGA2) antibodies to the surface of the liposomes
  • in the preparation of atosiban-conjugated liposomes
  • to prepare dual complementary liposome

Biochem/physiol Actions

DSPE-PEG(2000) carboxylic acid increases the blood circulation time of liposomes. The carboxyl group of DSPE-PEG(2000) carboxylic acid aids in covalent conjugation with antibodies.

Packaging

20 mL Clear Glass Screw Cap Vial (880135P-500mg)
5 mL Amber Glass Screw Cap Vial (880135P-100mg)
5 mL Amber Glass Screw Cap Vial (880135P-10mg)
5 mL Amber Glass Screw Cap Vial (880135P-25mg)
5 mL Amber Glass Screw Cap Vial (880135P-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Mashreghi et al.
Nanoscale research letters, 15(1), 101-101 (2020-05-10)
In this study, we have surface-functionalized PEGylated-nanoliposomal doxorubicin (DOX) with anti-EpCAM (epithelial cell adhesion molecule) aptamer via post-insertion of anti-EpCAM aptamer-conjugated DSPE-mPEG2000 into Caelyx® (ED-lip). The size, charge, release profile, and cytotoxicity and cellular uptake of formulation were determined. The
Carboxylic Acid: Key Role in Life Sciences (2019)
ITGA2 as a potential nanotherapeutic target for glioblastoma
Guo P, et al.
Scientific reports, 9(1), 6195-6195 (2019)
Junxue An et al.
Journal of colloid and interface science, 577, 92-100 (2020-05-31)
Phospholipids constitute biocompatible and safe excipients for pulmonary drug delivery. They can retard the drug release and, when PEGylated, also prolong the residence time in the lung. The aim of this work was to assess the structure and coherence of
Feifei Yang et al.
Small (Weinheim an der Bergstrasse, Germany), 16(7), e1906360-e1906360 (2020-01-24)
Hepatotoxicity is a key concern in the clinical translation of nanotherapeutics because preclinical studies have consistently shown that nanotherapeutics accumulates extensively in the liver. However, clinical-stage nanotherapeutics have not shown increased hepatotoxicity. Factors that can contribute to the hepatotoxicity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service