Skip to Content
Merck
All Photos(1)

Key Documents

860549P

Avanti

C24:1 Glucosyl(β) Ceramide (d18:1/24:1(15Z))

Avanti Research - A Croda Brand 860549P, powder

Synonym(s):

D-glucosyl-β-1,1′-N-nervonoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H91NO8
CAS Number:
Molecular Weight:
810.24
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860549P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860549P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O

InChI

1S/C48H91NO8/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-44(52)49-41(40-56-48-47(55)46(54)45(53)43(39-50)57-48)42(51)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2/h17-18,35,37,41-43,45-48,50-51,53-55H,3-16,19-34,36,38-40H2,1-2H3,(H,49,52)/

InChI key

WBOZIXHPUPAOIA-JZZPSRGZSA-N

General description

β-Glucosylceramides or glucocerebrosides (GlcCers) are synthesized from the glycosphingolipids by β-GlcCer synthase. They are synthesized in Golgi and endoplasmic reticulum and localized in the cytosolic side of cell membrane. GlcCers are omnipresent.

Application

C24:1 Glucosyl(β) Ceramide (d18:1/24:1(15Z)) may be used:
  • as a standard hexosylceramide for hydrophilic interaction chromatography-based liquid chromatography mass spectroscopy
  • to test its nuclear factor of activated T-cells (NFAT) activation
  • in human natural killer T (NKT) cell potency assays

Biochem/physiol Actions

β-Glucosylceramides (β-GlcCer) levels are elevated inlevels in Gaucher′s disease, due to the deficiency of β-glucosidase. β-GlcCer participates in the inflammatory and acquired immune responses by binding mincle on myeloid cells and has immunostimulatory activity.

Packaging

5 mL Amber Glass Screw Cap Vial (860549P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potent neutralizing anti-CD1d antibody reduces lung cytokine release in primate asthma model
Nambiar J, et al.
MAbs, 7(3), 638-650 (2015)
3-O-sulfo-beta-d-galactose moiety of endogenous sulfoglycolipids is a potential ligand for immunoglobulin-like receptor LMIR5
Phongsisay V, et al.
Molecular Immunology, 63(2), 595-599 (2015)
Diastereomer-specific quantification of bioactive hexosylceramides from bacteria and mammals
Von Gerichten J, et al.
Journal of Lipid Research, 58(6), 1247-1258 (2017)
Intracellular metabolite beta-glucosylceramide is an endogenous Mincle ligand possessing immunostimulatory activity
Nagata M, et al.
Proceedings of the National Academy of Sciences of the USA, 114(16), E3285-E3294 (2017)
Characterization and direct quantitation of cerebroside molecular species from lipid extracts by shotgun lipidomics
Han X and Cheng H
Journal of Lipid Research, 46(1), 163-175 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service