Skip to Content
Merck
All Photos(1)

Key Documents

Z768456

Removable Tough-Tags

L × W 1.5 in. × 0.50 in.

Synonym(s):

solvent resistant labels, temperature resistant labels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
55121611
NACRES:
NB.35

material

polyester
white

packaging

pkg of 1 roll (1000 labels)

manufacturer/tradename

Diversified Biotech RVHD-1000

L × W

1.5 in. × 0.50 in.

color

white

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ideal for use on reusable Labware such as beakers, bottles, flasks, glass vials, tubes, tube racks and pipettes. Enables sample tracking from purification to processing.

  • No residue - Easily remove from surfaces without leaving any adhesive
  • Track samples - Use labels to follow samples from purification to processing
  • Resistant - Resistant to most laboratory solvents and have shown short-term resistance to Acetone and Xylene
  • Useful - Great for beakers, bottles, flasks, glass vials, tubes, tube racks and pipettes

Legal Information

Tough-Tags is a trademark of Diversified Biotech

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service