Key Documents
Z742483
Prefilled tubes for BeadBlaster 24 Microtube and BeadBug homogenizer
2 ml tubes, with mixed format: 0.1 mm silica, 1.4 mm zirconium, and 4 mm silica
About This Item
UNSPSC Code:
41121800
NACRES:
NB.61
feature
2 ml tubes
packaging
pkg of 50
manufacturer/tradename
Benchmark Scientific D1034-MX
Related Categories
General description
Prefilled tubes; Compatible with all common bead homogenizers
Legal Information
BeadBlaster is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)