Key Documents
Z282049
Coors alumina pestle
for Z281999
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
material
alumina
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 60359
pestle L
57 mm
Related Categories
General description
99.5% alumina. Extremely hard and completely nonabsorbent surface. Order mortar separately.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)