Key Documents
Z231312
Sigma-Aldrich
Giant-size molecular model kit
basic kit
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
description
no. of flexible bonds 16
no. of rigid bonds 50
feature
no. of atoms 32
General description
Large, lightweight molecular models for classroom demonstration of organic and inorganic structures. For building models to scale of 20 cm/Å and 8 cm/Å. Bond lengths range from 5.5 to 27 cm. Atomic centers are 5 cm in diameter, finished in standard atom colors, and have a reinforced vinyl receptor at each bond position for insertion of connecting rods (bonds). All model kits include storage box and instructions. Additional atoms and bonds are available separately.