Key Documents
Z215937
Solid PTFE stopper
joint: ST/NS 29/42
Synonym(s):
flask stopper, lab stopper, stoppers
About This Item
UNSPSC Code:
41122500
NACRES:
NB.35
Pricing and availability is not currently available.
Joint
joint: ST/NS29/42
manufacturer/tradename
Cowie 010.129
General description
Machined from solid PTFE with trapezoidal seals for medium to high vacuum performance up to 280°C (540°F).
Choose from one of the most recent versions:
Certificates of Analysis (COA)