As listed in the 'PROPERTIES' section, under 'fittings', the external valve is CGA 180 male and the internal is CGA 110 female. A CGA 110 male fitting would be required to cap this valve for the inner thread. The opposite is true for the external CGA 180 male thread; a CGA 180 female fitting would be required to cap the exterior valve.
Z175552
Lecture-bottle valve
CGA Inlet for 180M/110F, brass
Synonym(s):
Lecture Bottle Control Valve
About This Item
Recommended Products
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
-
What size cap or plug should be used for this valve?
1 answer-
Helpful?
-