Z175013
Adapter for ACE Micro/Mini-Lab® kit
Mini adapter
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
joint ST/NS14/10
manufacturer/tradename
Ace Glass 502825
Looking for similar products? Visit Product Comparison Guide
General description
With threaded nylon bushing to accommodate 6.5 to 7 mm diam. thermometer or tubing.
Legal Information
Micro/Mini-Lab is a registered trademark of Ace Glass, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Epidemiology and infection, 143(9), 1833-1838 (2014-11-02)
No published studies have discussed details of the prognosis and survival of patients with severe avian influenza A(H7N9) infection. In this study we analysed 128 laboratory-confirmed cases of severe H7N9 infection in Zhejiang province, the most affected region during the
Molecular biology and evolution, 32(8), 2060-2071 (2015-04-11)
The majority of human group A rotaviruses possess the P[8] VP4 genotype. Recently, a genetically distinct subtype of the P[8] genotype, also known as OP354-like P[8] or lineage P[8]-4, emerged in several countries. However, it is unclear for how long
Transfusion, 55(6 Pt 2), 1407-1410 (2014-11-22)
The rare amorph Rhnull phenotype is caused by silent alleles at the RH locus and usually arises in consanguineous families. To date, only five molecular backgrounds have been identified in five unrelated families. Subjects with Rhnull red blood cells (RBCs)
Oncotarget, 6(15), 13176-13200 (2015-05-12)
SKBR3-cells, characterized by ERBB2/RARA co-amplification, represent a subgroup of HER2+ breast-cancers sensitive to all-trans retinoic acid (ATRA) and Lapatinib. In this model, the two agents alone or in combination modulate the expression of 174 microRNAs (miRs). These miRs and predicted
Pregnancy hypertension, 5(4), 294-297 (2015-11-26)
There is recent interest for uniform placental collection protocols so laboratories can share samples. However, concerns have been raised a wash step at collection causes significant loss of sFlt-1 and soluble endoglin, among the most studied proteins in placentology. We
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service