Z124400
Natural rubber septa
red rubber, mixed set (145 pieces with carrying box)
All Photos(1)
About This Item
UNSPSC Code:
41121800
eCl@ss:
32119211
NACRES:
NB.35
Recommended Products
material
red rubber
Related Categories
General description
Mixed set contains:
25 each of the septa for 5 mm NMR tubes; 7 mm O.D. glass tubing; and 8 or 9 mm O.D. glass tubing
20 each of the septa for 9 or 10 mm O.D. glass tubing; and 10/30 joints and small test tubes
10 each of the septa for 14/20 joints; 19/22 joints; and 24/40 joints
25 each of the septa for 5 mm NMR tubes; 7 mm O.D. glass tubing; and 8 or 9 mm O.D. glass tubing
20 each of the septa for 9 or 10 mm O.D. glass tubing; and 10/30 joints and small test tubes
10 each of the septa for 14/20 joints; 19/22 joints; and 24/40 joints
Kit Components Also Available Separately
Product No.
Description
SDS
- Z564710Sleeve stopper septa, bottom I.D. × O.D. 9.3 mm × 15.7 mm, red 2 EASDS
- Z565644Sleeve stopper septa, bottom I.D. × O.D. 1.5 mm × 3.9 mm, red 5 EASDS
- Z565660Sleeve stopper septa, bottom I.D. × O.D. 2.2 mm × 4.5 mm, red 5 EASDS
- Z565687Sleeve stopper septa, bottom I.D. × O.D. 2.4 mm × 5.3 mm, red 5 EASDS
- Z565709Sleeve stopper septa, bottom I.D. × O.D. 3.4 mm × 6.6 mm, red 4 EASDS
- Z564699Sleeve stopper septa, bottom I.D. × O.D. 7.9 mm × 14 mm, red 2 eaSDS
- Z564699Sleeve stopper septa, bottom I.D. × O.D. 7.9 mm × 14 mm, redSDS
- Z564737Sleeve stopper septa, bottom I.D. × O.D. 12.7 mm × 23.7 mm, red 2 EASDS
See All (8)
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)