Skip to Content
Merck
All Photos(1)

Key Documents

Z102571

Sigma-Aldrich

Aldrich® jacketed Dean-Stark trap

capacity 325 mL, joint: ST/NS 24/40

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104812
NACRES:
NB.43

Joint

joint: ST/NS24/40

capacity

325 mL

General description

For azeotroping large amounts of water, 325 mL capacity. With 24/40 joints and 4 mm bore PTFE stopcock.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service