WHA10409834
Whatman® mixed cellulose ester ME 26 filter discs 0.6 μm pore size
black, hydrophilic, white grid (3.1 mm), sterile, for membrane butler, 400 EA, 400 ea, 50 mm diam
Synonym(s):
Z750549
About This Item
product name
Whatman® ME membranes for microbiological control, ME26/31, 3.1 mm grid, for Membrane-Butler, pore size 0.6 μm, white grid, diam. 50 mm
material
white grid
sterility
sterile
packaging
pk of 100
manufacturer/tradename
Whatman 10409834
Whatman Article No. 28415264 (US reference)
diam.
50 mm
membrane diam.
47 mm
pore size
0.6 μm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Sterile options available for critical applications.
- Excellent contrast for easier particle detection.
- Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity.
- Black plain and black gridded membranes have a mix of cellulose nitrate and cellulose acetate.
- The membrane offers a high degree of internal surface area for greater adsorption of product.
- Higher dirt loading capacity.
- Biologically inert with good thermal stability.
- No surfactants to contaminate samples.
- Uniform microporous structure of membrane gives high flow rates.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service