WHA10401731
Whatman® mixed cellulose ester ME 24 filter discs 0.2 μm pore size
white (no grid), hydrophilic, nonsterile, interleaving paper, 25 EA, 25 ea, 142 mm diam
Synonym(s):
Z697354, cellulose membrane, membrane filter, millipore filter
About This Item
Recommended Products
Product Name
Whatman® membrane filters mixed cellulose ester, pore size 0.2 μm, diam. 142 mm
sterility
non-sterile
feature
hydrophilic
packaging
pkg of 25 ea
manufacturer/tradename
Whatman 10401731
Whatman Article No. 28415048 (US reference)
diam.
142 mm
pore size
0.2 μm
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Plain and gridded options for ease of colony and cell counting methods
- Supports visual enumeration of bacteria
- Appropriate for bacterial culturing, including E. coli culturing
- Biologically inert with good thermal stability
- Uniform microporous structure results in high flow rates when used for filtering
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service