Skip to Content
Merck
All Photos(1)

Key Documents

W515604

Sigma-Aldrich

Isopropyl palmitate

≥90%

Synonym(s):

Isopropyl hexadecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOCH(CH3)2
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786567
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.606
NACRES:
NA.21
grade:
Fragrance grade
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

Fragrance grade
Halal

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥90%

refractive index

n20/D 1.438 (lit.)

mp

11-13 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

oily

SMILES string

CCCCCCCCCCCCCCCC(=O)OC(C)C

InChI

1S/C19H38O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19(20)21-18(2)3/h18H,4-17H2,1-3H3

InChI key

XUGNVMKQXJXZCD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Isopropyl palmitate is an aliphatic ester used as a flavoring ingredient in food industry. It is one of the volatile compounds found in Psidium salutare fruits and boiled buckwheat flour.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Burdock GA
Encyclopedia of Food and Color Additives, 1, 1496-1496 (1997)
Volatile flavor compounds of boiled buckwheat flour
Yajima I, et al.
Agricultural and Biological Chemistry, 47(4), 729-738 (1983)
Volatile compounds of Psidium salutare (HBK) Berg. fruit.
Pino JA, et al.
Journal of Agricultural and Food Chemistry, 50(18), 5146-5148 (2002)
Varaporn Buraphacheep Junyaprasert et al.
Drug delivery, 15(5), 323-330 (2008-09-03)
The skin permeation enhancement of many kinds of drugs and cosmetic substances by microemulsions has been widely known; however, the correlations between microemulsion microstructures and the efficiency of skin permeation are not fully elucidated. Therefore, the aim of our study
N Pfammatter et al.
Biochemical and biophysical research communications, 161(3), 1244-1251 (1989-06-30)
Baker yeast cells are solubilized in organic solvents by the use of surfactants and small amounts of water. Data are reported for three different systems, Tween/Isopropylpalmitat, Asolectin/IPP, Asolectin/Hexadecane. The viability (life-capability) can remain as high as 80% for 10 Days

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service