Skip to Content
Merck
All Photos(1)

Key Documents

W397301

Sigma-Aldrich

2-Methyl-3-furanthiol acetate

98%, FG

Synonym(s):

2-Methyl-3-furyl thioacetate, 2-Methylfuran-3-thioacetate, 3-(Acetylthio)-2-methylfuran, S-(2-methyl-3-furyl) ethanethioate, Ethanethioic acid, S-(2-methyl-3-furanyl) ester, MFT Acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2S
CAS Number:
Molecular Weight:
156.20
FEMA Number:
3973
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.153
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

98%

refractive index

n20/D 1.520

bp

225-235 °C/29.9 mmHg

density

1.138 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty

SMILES string

CC(=O)Sc1ccoc1C

InChI

1S/C7H8O2S/c1-5-7(3-4-9-5)10-6(2)8/h3-4H,1-2H3

InChI key

PQFIBPDAGFGLBY-UHFFFAOYSA-N

General description

2-Methyl-3-furanthiol acetate is a volatile sulfur-containing furan compound present as a flavor compound in Scheurebe wines and purple passion fruit.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile compounds affecting the aroma of Vitis vinifera L. cv. Scheurebe.
Krammer GE, et al.
ACS Symp. Ser., 714, 53?65-53?65 (1998)
Determination and isolation of a thioesterase from passion fruit (Passiflora edulis Sims) that hydrolyzes volatile thioesters.
Tapp EJ, et al.
Journal of Agricultural and Food Chemistry, 56(15), 6623-6630 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service