Skip to Content
Merck
All Photos(1)

Documents

W334308

Sigma-Aldrich

Ethyl 3-(methylthio)propionate

≥99%, FCC, FG

Synonym(s):

Ethyl 3-(methyl thio) propanoate, Ethyl methyl mercaptopropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CO2C2H5
CAS Number:
Molecular Weight:
148.22
FEMA Number:
3343
EC Number:
Council of Europe no.:
11476
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.053
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥99%

refractive index

n20/D 1.46 (lit.)

bp

197 °C (lit.)

density

1.032 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; pineapple; sulfurous

SMILES string

CCOC(=O)CCSC

InChI

1S/C6H12O2S/c1-3-8-6(7)4-5-9-2/h3-5H2,1-2H3

InChI key

YSNWHRKJEKWJNY-UHFFFAOYSA-N

Application


  • Identification of new strawberry sulfur volatiles and changes during maturation.: Du et al. explore sulfur volatiles in strawberries, noting the presence of ethyl 3-(methylthio)propionate among new compounds discovered. The study provides insights into how these volatiles evolve during fruit maturation, impacting flavor and aroma profiles (Du et al., 2011).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justyna Aszyk et al.
Journal of chromatography. A, 1547, 86-98 (2018-03-15)
Many flavour compounds that are present in e-liquids for e-cigarettes are responsible for specific tastes and smoking sensations for users. Data concerning content and specific types of flavours is often limited and unknown to users. The aim of the research
Justyna Aszyk et al.
Analytica chimica acta, 1038, 67-78 (2018-10-04)
Currently, there is lack of standardized conditions for the collection and analysis of e-cigarette (EC) aerosol. Considering the urgent need for the development of these guidelines, a procedure for EC aerosol analysis was developed. A novel automatic e-cigarette aerosol generator
Urszula Dziekońska-Kubczak et al.
Molecules (Basel, Switzerland), 25(5) (2020-03-19)
Volatile sulfur compounds (VSCs) play an important role in the aroma profile of fermented beverages. However, because of their low concentration in samples, their analysis is difficult. The headspace solid-phase microextraction (HS-SPME) technique coupled with gas chromatography and mass spectrometry
Shuang Chen et al.
Journal of food science, 82(12), 2816-2822 (2017-11-14)
This study investigated the aroma contribution of volatile sulfur compounds (VSCs) in Moutai liquors. The VSCs were analyzed using headspace solid-phase microextraction-gas chromatography-pulsed flame photometric detection (HS-SPME-GC-PFPD). The influences of SPME fibers, ethanol content in the sample, pre-incubation time, and
Sha Sha et al.
Journal of agricultural and food chemistry, 65(1), 123-131 (2016-12-19)
The aroma profile of Chinese roasted sesame-like flavor type liquor was investigated by means of headspace solid phase microextraction-aroma extract dilution analysis (HS-SPME-AEDA). Sixty-three odor-active regions were detected by HS-SPME-AEDA with flavor dilution (FD) factors >5, and 58 of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service