W317713
p-Mentha-8-thiol-3-one
mixture of cis and trans, natural, ≥95%, FG
Synonym(s):
8-Mercaptomenthone, buchu mercaptan, 2-(1-Mercapto-1-methylethyl)-5-methylcyclohexanone
About This Item
Fragrance grade
Halal
Kosher
natural
Recommended Products
grade
FG
Fragrance grade
Halal
Kosher
natural
Quality Level
300
400
Agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
Assay
≥95%
optical activity
[α]20/D −21°, neat
composition
may contain trace Pulegone
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
refractive index
n20/D 1.495 (lit.)
bp
56 °C/0.1 mmHg (lit.)
density
1 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
greener alternative category
Organoleptic
berry; tropical; minty; fruity; sulfurous
SMILES string
CC1CCC(C(=O)C1)C(C)(C)S
InChI
1S/C10H18OS/c1-7-4-5-8(9(11)6-7)10(2,3)12/h7-8,12H,4-6H2,1-3H3
InChI key
RVOKNSFEAOYULQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- RIFM fragrance ingredient safety assessment, p-mentha-8-thiol-3-one, CAS Registry Number 38462-22-5.: This review provides a comprehensive safety assessment of p-Mentha-8-thiol-3-one, focusing on its applications in fragrance formulations. The study evaluates the compound′s toxicological profile and discusses its safe usage levels in consumer products. The research highlights the compound′s potential as a fragrance ingredient due to its unique olfactory properties. (Api et al., 2021).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
226.4 °F - closed cup
Flash Point(C)
108 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service