Skip to Content
Merck
All Photos(1)

Key Documents

W317403

Sigma-Aldrich

4-Hydroxy-2,5-dimethyl-3(2H)-furanone

≥98%, FCC, FG

Synonym(s):

2,5-Dimethyl-4-hydroxy-3(2H)-furanone, Furaneol, Strawberry furanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
FEMA Number:
3174
Beilstein:
1281357
EC Number:
Council of Europe no.:
536
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.010
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥98%

mp

73-77 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

caramel; strawberry; sweet

storage temp.

2-8°C

SMILES string

CC1OC(C)=C(O)C1=O

InChI

1S/C6H8O3/c1-3-5(7)6(8)4(2)9-3/h3,8H,1-2H3

InChI key

INAXVXBDKKUCGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Hydroxy-2,5-dimethyl-3(2H)-furanone is a flavor compound that occurs in fruits such as pineapple, strawberry and mango.

Application


  • FaQR, required for the biosynthesis of the strawberry flavor compound 4-hydroxy-2,5-dimethyl-3(2H)-furanone, encodes an enone oxidoreductase.: This study identifies the FaQR gene as essential for the biosynthesis of the key strawberry flavor compound, 4-hydroxy-2,5-dimethyl-3(2H)-furanone. The research highlights the enzymatic pathway involved and its potential applications in biotechnology for flavor enhancement in food products (Raab et al., 2006).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rational synthesis of 4-hydroxy-2, 5-dimethyl-3 (2H)-furanone, a flavor component of pineapple
Henry DW & Silverstein RM.
The Journal of Organic Chemistry, 31(7), 2391-2394 (1966)
Estimation of 2, 5?dimethyl?4?hydroxy?3 (2H)?furanone (FURANEOLA?) in cultivated and wild strawberries, pineapples and mangoes
Pickenhagen W, et al.
Journal of the Science of Food and Agriculture, 32(11), 1132-1134 (1981)
Thomas Raab et al.
The Plant cell, 18(4), 1023-1037 (2006-03-07)
The flavor of strawberry (Fragaria x ananassa) fruit is dominated by an uncommon group of aroma compounds with a 2,5-dimethyl-3(H)-furanone structure. We report the characterization of an enzyme involved in the biosynthesis of 4-hydroxy-2,5-dimethyl-3(2H)-furanone (HDMF; Furaneol), the key flavor compound
Silke Illmann et al.
Journal of agricultural and food chemistry, 57(7), 2889-2895 (2009-03-05)
The formation of 4-hydroxy-2,5-dimethyl-3(2H)-furanone (HDMF) was studied in aqueous model systems containing L-rhamnose and L-lysine. The approach consisted in systematically varying four reaction parameters (rhamnose concentration, rhamnose to lysine ratio, pH, and phosphate concentration) at 3 levels. A fractional factorial
Woo Sang Sung et al.
Life sciences, 80(6), 586-591 (2006-11-14)
2,5-Dimethyl-4-hydroxy-3(2H)-furanone (DMHF), an aroma compound found in a number of fruits and foods, has shown various biological properties in animal models, but its antimicrobial effect remains poorly understood. The current study investigated the antimicrobial effect of DMHF using human pathogenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service