Skip to Content
Merck
All Photos(1)

Key Documents

W317101

Sigma-Aldrich

cis-3-Hexenyl acetate

≥98%, stabilized, FCC, FG

Synonym(s):

(3Z)-3-Hexen-1-ol acetate, (3Z)-C-3-Hexenyl acetate, Leaf acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
142.20
FEMA Number:
3171
EC Number:
Council of Europe no.:
644
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.197
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥98%

contains

α-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.427 (lit.)

bp

75-76 °C/23 mmHg (lit.)

density

0.897 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fresh; green; fruity; sweet

SMILES string

[H]\C(CC)=C(/[H])CCOC(C)=O

InChI

1S/C8H14O2/c1-3-4-5-6-7-10-8(2)9/h4-5H,3,6-7H2,1-2H3/b5-4-

InChI key

NPFVOOAXDOBMCE-PLNGDYQASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

cis-3-Hexenyl acetate is a volatile aroma compound naturally found in virgin olive oil and Jasminum sambac flower. It is used as a flavoring agent in the food and cosmetics industries.

Application


  • Volatiles from cotton aphid (Aphis gossypii) infested plants attract the natural enemy Hippodamia variegata.: This study explored how volatiles, including cis-3-Hexenyl acetate, emitted by cotton plants infested with Aphis gossypii attract the natural predator Hippodamia variegata, highlighting its potential in biological pest control strategies (Yi et al., 2023).

Biochem/physiol Actions

Taste at 1-15 ppm

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F

Flash Point(C)

57 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of aroma-active compounds in Jasminum sambac concrete by aroma extract dilution analysis and odour activity value
Zhang J, et al.
Flavour and Fragrance Journal, 36(2), 197-206 (2021)
Guilherme G Teixeira et al.
Talanta, 226, 122122-122122 (2021-03-08)
An electronic nose, comprising nine metal oxide sensors, has been built aiming to classify olive oils according to the fruity intensity commercial grade (ripely fruity or light, medium and intense greenly fruity), following the European regulated complementary terminology. The lab-made
Virgin olive oil odour notes: their relationships with volatile compounds from the lipoxygenase pathway and secoiridoid compounds
Angerosa F, et al.
Food Chemistry, 68(3), 283-287 (2000)
Jürgen Engelberth et al.
Molecular plant-microbe interactions : MPMI, 20(6), 707-716 (2007-06-09)
The induction of jasmonic acid (JA) is one of the major signaling events in plants in response to insect herbivore damage and leads to the activation of direct and indirect defensive measures. Green leafy volatiles, which constitute a major portion
Nasser Said Mandour et al.
Journal of chemical ecology, 37(3), 267-272 (2011-02-19)
Females of the gregarious endoparasitoid Cotesia kariyai were attracted to a blend of volatiles released from corn plants infested with larvae of their host, the common armyworm (Mythimna separata). We investigated the effects of time after the last infestation (1-168

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service