Skip to Content
Merck
All Photos(1)

Key Documents

W313203

Sigma-Aldrich

2-Isobutyl-3-methoxypyrazine

≥99%, FG

Synonym(s):

2-methoxy-3-(2-methylpropyl)pyrazine, 2-methoxy-3-isobutylpyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N2O
CAS Number:
Molecular Weight:
166.22
FEMA Number:
3132
EC Number:
Council of Europe no.:
11338
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.043
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥99%

refractive index

n20/D 1.49 (lit.)

density

0.99 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; vegetable; pepper

SMILES string

COc1nccnc1CC(C)C

InChI

1S/C9H14N2O/c1-7(2)6-8-9(12-3)11-5-4-10-8/h4-5,7H,6H2,1-3H3

InChI key

UXFSPRAGHGMRSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Isobutyl-3-methoxypyrazine is the main characteristic flavor/aroma constituent of bell peppers. It is also found in fresh jalapeno peppers and baked potato.

Application


  • RIFM Fragrance Ingredient Safety Assessment, 2-Isobutyl-3-methoxypyrazine, CAS Registry Number 24683-00-9.: This safety assessment reviews the toxicological data of 2-Isobutyl-3-methoxypyrazine, ensuring its safe use in fragrances (Api et al., 2024).

  • Source-Sink Manipulations Have Major Implications for Grapevine Berry and Wine Flavonoids and Aromas That Go Beyond the Changes in Berry Sugar Accumulation.: This article discusses the impact of 2-Isobutyl-3-methoxypyrazine on wine aroma, focusing on agricultural practices and their influence on grape quality (Martínez-Lüscher and Kurtural, 2023).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile components and pungency in fresh and processed Jalapeno peppers.
Huffman VL, et al.
Journal of Food Science, 43(6), 1809-1811 (1978)
Volatile flavour components of baked potato flesh. A comparison of eleven potato cultivars.
Duckham SC, et al.
Molecular Nutrition And Food Research, 45(5), 317-323 (2001)
Katja Suklje et al.
Journal of agricultural and food chemistry, 60(37), 9454-9461 (2012-09-06)
Grape berries were classified according to diameter and total soluble solids (TSS) to study the effect of light and temperature on methoxypyrazines (MPs), glutathione (GSH), and hydroxycinnamates (HCAs) during the ripening of Sauvignon blanc. The light exposure of the fruiting
Philippus Alberts et al.
Journal of agricultural and food chemistry, 57(20), 9347-9355 (2009-10-08)
An LC-MS/MS method for the trace-level determination of 3-alkyl-2-methoxypyrazines in Sauvignon blanc wines is described. 3-Isobutyl-2-methoxypyrazine (IBMP), 3-isopropyl-2-methoxypyrazine (IPMP) and 3-sec-butyl-2-methoxypyrazine (SBMP) were analyzed by reversed phase liquid chromatography coupled to atmospheric pressure chemical ionization, as electrospray ionization was found
Maria Staiano et al.
Biopolymers, 89(4), 284-291 (2007-12-22)
The effects of hydrostatic pressure on the structure and stability of porcine odorant-binding protein (pOBP) in the presence and absence of the odorant molecule 2-isobutyl-3-methoxypyrazine (IBMP) were studied by steady-state and time-resolved fluorescence spectroscopy as well as by molecular dynamics

Protocols

Separation of 2-Isopropyl-3-methoxypyrazine 20 ppt; 2-Isobutyl-3-methoxypyrazine; 2-Methylisoborneol; (±)-Geosmin 10 ppt

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service