Skip to Content
Merck
All Photos(1)

Key Documents

W287407

Sigma-Aldrich

Phenylacetaldehyde

≥95%, FCC, FG

Synonym(s):

2-Phenylacetaldehyde, α-Tolyaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CHO
CAS Number:
Molecular Weight:
120.15
FEMA Number:
2874
Beilstein:
385791
EC Number:
Council of Europe no.:
116
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39023706
PubChem Substance ID:
Flavis number:
5.030
NACRES:
NA.21
Organoleptic:
chocolate; green; honey; hyacinth
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

description

may contain Citric acid

Assay

≥95%

refractive index

n20/D 1.535 (lit.)

bp

195 °C

mp

−10 °C (lit.)

density

1.035 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

chocolate; green; honey; hyacinth

storage temp.

2-8°C

SMILES string

O=CCc1ccccc1

InChI

1S/C8H8O/c9-7-6-8-4-2-1-3-5-8/h1-5,7H,6H2

InChI key

DTUQWGWMVIHBKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenylacetaldehyde is an important aroma volatile found in tomato and roses. It has also been identified in potato, roasted cocoa beans and honey. Phenylacetaldehyde is also a potent moth attractant.

Biochem/physiol Actions

Taste at 5.0 ppm

Other Notes

Natural occurrence: Apple, apricot, bilbery, cherry, grapefruit, guava, orange peel, peach, raisin, grape, asparagus, blackberry, papaya, melon, cabbage, sweet pepper and celery leaves.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

154.4 °F - (External MSDS)

Flash Point(C)

68 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenylacetaldehyde: a chemical attractant for common green lacewings (Chrysoperla carnea sl, Neuroptera: Chrysopidae).
Toth MIKLOS, et al
European Journal of Entomology, 103(1), 267-267 (2006)
A review of volatile analytical methods for determining the botanical origin of honey.
Cuevas-Glory LF, et al.
Food Chemistry, 103(3), 1032-1043 (2007)
Tomato phenylacetaldehyde reductases catalyze the last step in the synthesis of the aroma volatile 2-phenylethanol.
Tieman DM, et al.
Phytochemistry, 68(21), 2660-2669 (2007)
Phenylacetaldehyde attracts moths to bladder flower and to blacklight traps.
Cantelo WW and Jacobson M.
Environmental Entomology, 8(3), 444-447 (1979)
Characterization of some volatile potato components.
Buttery RG, et al.
Journal of Agricultural and Food Chemistry, 18(3), 538-539 (1970)

Protocols

GC Analysis of Aldehydes (PFBOA Derivatives) in Beer on SLB®-5ms after SPME using a 65 µm PDMS/DVB Fiber

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service