Skip to Content
Merck
All Photos(1)

Key Documents

W284815

Sigma-Aldrich

Peppermint oil

natural, FCC, FG

Synonym(s):

Mentha piperita

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2848
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
biological source:
Mentha piperita L.
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

Mentha piperita L.

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.230
FDA 21 CFR 182.20

optical activity

[α]20/D −26°, neat

origin

USA origin

refractive index

n20/D 1.461 (lit.)

bp

215 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

peppermint

Looking for similar products? Visit Product Comparison Guide

Application


  • A full factorial experimental design to study the effect of flavoring agents on the mechanical properties of curcumin chewing gum tablets with high solids content.: The research explores how different flavoring agents, including peppermint oil, affect the mechanical properties of curcumin chewing gum tablets. This study is relevant for developing optimized formulations in pharmaceutical biochemistry (Al Hagbani et al., 2020).

  • Stress Response and Virulence Potential Modulating Effect of Peppermint Essential Oil in Campylobacter jejuni.: This investigation looks into how peppermint essential oil modulates stress responses and virulence in Campylobacter jejuni, highlighting its potential application in biochemical studies related to microbial pathogenesis and infection control (Kovács et al., 2019).

  • Application of in vitro lipolysis for the development of oral self-emulsified delivery system of nimodipine.: This paper discusses the use of in vitro lipolysis to develop an oral self-emulsified delivery system for nimodipine, incorporating peppermint oil as a key component. It emphasizes the role of biochemistry in enhancing drug delivery methods (Alayoubi et al., 2018).

Preparation Note

Extraction method: steam distillation

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Alam et al.
Mymensingh medical journal : MMJ, 22(1), 27-30 (2013-02-19)
Irritable bowel syndrome (IBS) is one of the most common functional gastrointestinal disorder which is associated with considerable sufferings of patient and Peppermint oil is volatile oil, its active principle is menthol-contain a cyclic monoterpine which has anti-spasmotic properties due
Naoki Hiki et al.
Digestive endoscopy : official journal of the Japan Gastroenterological Endoscopy Society, 24(2), 79-86 (2012-02-22)
Peppermint oil solution was found to be effective for reducing gastric spasm during upper gastrointestinal endoscopy. The aim of the present study was to assess whether the gastric peristalsis-suppressing effect is dose-dependently induced by L-menthol, the major constituent of peppermint
Atsushi Imagawa et al.
Digestive diseases and sciences, 57(9), 2379-2384 (2012-05-09)
Although hyoscine butyl bromide (HB) and glucagon (GL) are often used as antispasmodic drugs during esophagogastroduodenoscopy (EGD), these agents may cause adverse effects. Recently, it was reported that peppermint oil solution (PO) was very effective and had few side effects.
Sonia Hines et al.
The Cochrane database of systematic reviews, 4(4), CD007598-CD007598 (2012-04-20)
Postoperative nausea and vomiting is a common and unpleasant phenomenon and current therapies are not always effective for all patients. Aromatherapy has been suggested as a possible addition to the available treatment strategies. This review sought to establish what effect
Katharina Heimes et al.
Phytotherapy research : PTR, 25(5), 702-708 (2010-11-16)
Peppermint oil (Mentha × piperita L. (Lamiaceae) has been shown to exert potent antiemetic properties, but its mode of action has not yet been elucidated. Among its active constituents (-)-menthol is the most important. Three different in vitro models were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service