Skip to Content
Merck
All Photos(1)

Key Documents

W269301

Sigma-Aldrich

Methyl butyrate

≥98%, FG

Synonym(s):

Butanoic acid methyl ester, Methyl butanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOCH3
CAS Number:
Molecular Weight:
102.13
FEMA Number:
2693
Beilstein:
1740743
Council of Europe no.:
263
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.038
NACRES:
NA.21
Organoleptic:
apple; ethereal
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
(EU Regulation 1223/2009)
(follows IFRA guidelines)
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

(EU Regulation 1223/2009)
(follows IFRA guidelines)
meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 30 °C)

Assay

≥98%

expl. lim.

3.5 %

refractive index

n20/D 1.385 (lit.)

bp

102-103 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; ethereal

SMILES string

CCCC(=O)OC

InChI

1S/C5H10O2/c1-3-4-5(6)7-2/h3-4H2,1-2H3

InChI key

UUIQMZJEGPQKFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.0 °F

Flash Point(C)

11.67 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C D Hull et al.
Journal of chemical ecology, 27(5), 889-906 (2001-07-27)
Single-unit electrophysiology was used to record the nerve impulses from the carbon dioxide receptors of female Queensland fruit flies, Bactroera tryoni. The receptors responded to stimulation in a phasic-tonic manner and also had a period of inhibition of the nerve
A Nakagawa et al.
Journal of biochemistry, 96(3), 815-820 (1984-09-01)
Purified Pseudomonas fluorescens lipase [EC 3.1.1.3] exhibited slight activity on water-soluble esters such as methyl butyrate, and this activity was increased on addition of Brij 58 (20 oxyethylene hexadecyl ether) to the solution. This stimulating effect of Brij 58 on
S H Sterri et al.
Biochemical pharmacology, 34(15), 2779-2785 (1985-08-01)
A radiochemical assay for carboxylesterase based on the substrate methyl[1-14C]butyrate is described. The blank value corresponds to 1.04 micrograms (liver)-1.44 mg (plasma) of tissues with the highest and lowest activity respectively, which constitute the sensitivity of the method. The hydrolysis
D Roy et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 15(17), 3839-3847 (2014-09-10)
An easily dispersible multiwalled carbon nanotube (MWCNT) derivative is prepared, and provides a platform for the synthesis of the phenyl butyric acid methyl ester (PCBM) analog. The carbene addition reaction of MWCNTs makes derivatives that are less soluble in organic
Guoyi Ma et al.
Cancer chemotherapy and pharmacology, 63(4), 673-680 (2008-07-04)
Breast cancer is one of the most prevalent woman cancers. Genomic instability, accumulative mutations, and subsequent changes in intracellular signaling cascades play key roles in the development of human breast cancers. Activation of nuclear factor-kappaB (NF-kappaB) has been implicated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service