Skip to Content
Merck
All Photos(1)

Documents

W221503

Sigma-Aldrich

Butyl sulfide

≥95%, FG

Synonym(s):

Dibutyl sulfide, 1,1′-Thiobisbutane, Butyl sulfide, NSC 8460

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3S(CH2)3CH3
CAS Number:
Molecular Weight:
146.29
FEMA Number:
2215
Beilstein:
1732829
EC Number:
Council of Europe no.:
484
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.007
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

5.07 (vs air)

vapor pressure

5.17 mmHg ( 37.7 °C)

Assay

≥95%

refractive index

n20/D 1.452 (lit.)

bp

188-189 °C (lit.)

mp

−76 °C (lit.)

density

0.838 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; sulfurous

SMILES string

CCCCSCCCC

InChI

1S/C8H18S/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3

InChI key

HTIRHQRTDBPHNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl sulfide is a volatile sulfur compound that can be used as a flavoring agent.

Application


  • Room-Temperature Synthesis of Thioether-Stabilized Ruthenium Nanocubes and Their Optical Properties.: This article presents a method for synthesizing thioether-stabilized ruthenium nanocubes at room temperature, using butyl sulfide as a stabilizing agent. The study details the optical properties of the nanocubes, which could have implications for their use in catalysis and materials science (Adams et al., 2023).

Biochem/physiol Actions

Taste at 5 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Burdock GA
Encyclopedia of Food and Color Additives, 1, 356-357 (1997)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 356-357 (1997)
Stanisław Popiel et al.
Journal of hazardous materials, 157(2-3), 319-327 (2008-02-23)
A scheme of dibutyl sulfide (DBS) oxidation with ozone and generation of transitional products was determined in this study. The main identified intermediate product was dibutyl sulfoxide (DBSO), and the main end product of DBS oxidation was dibutyl sulfone (DBSO2).
Izabela Sobczak et al.
Journal of hazardous materials, 179(1-3), 444-452 (2010-04-07)
Zeolites exhibiting different structures (Y, Beta, and ZSM-5) were modified with gold and iron and applied for odour adsorption from the air containing dibutyl sulphide (Bu(2)S) used as a representative odour producing compound. The structure of the zeolites used determines
Ashleigh E Baber et al.
ACS nano, 2(11), 2385-2391 (2009-02-12)
This paper describes a fundamental, single-molecule study of the motion of a set of thioethers supported on Au surfaces. Thioethers constitute a simple, robust system with which molecular rotation can be actuated both thermally and mechanically. Low-temperature scanning tunneling microscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service