Skip to Content
Merck
All Photos(1)

Key Documents

W208019

Sigma-Aldrich

Isoamyl octanoate

natural, ≥98%, FG

Synonym(s):

Isoamyl caprylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CO2CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
214.34
FEMA Number:
2080
EC Number:
Council of Europe no.:
401
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.120
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

description

mixture of isoamyl octanoate and 2-methylbutyl octanoate

Assay

≥98%

refractive index

n20/D 1.426 (lit.)

bp

267-268 °C

density

0.861 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coconut; green; waxy; fruity; pineapple; sweet

SMILES string

CCCCCCCC(=O)OCCC(C)C

InChI

1S/C13H26O2/c1-4-5-6-7-8-9-13(14)15-11-10-12(2)3/h12H,4-11H2,1-3H3

InChI key

XKWSWANXMRXDES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Natural occurrence: Scotch, whiskey, bourbon whiskey, wine, banana, apple and strawberry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service