Skip to Content
Merck
All Photos(1)

Key Documents

T6601

Sigma-Aldrich

3,4,5,6-Tetrachloro-1,2-benzoquinone

97%

Synonym(s):

o-Chloranil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6Cl4(=O)2
CAS Number:
Molecular Weight:
245.88
Beilstein:
1309782
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

126-129 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl

InChI

1S/C6Cl4O2/c7-1-2(8)4(10)6(12)5(11)3(1)9

InChI key

VRGCYEIGVVTZCC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank Wurche et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(11), 2707-2721 (2004-06-15)
The effect of pressure on the oxidation of hydroarenes 3-9 with 2,3-dichloro-5,6-dicyano-1,4-quinone (DDQ; 1 a) or o-chloranil (10), leading to the corresponding arenes, has been investigated. The activation volumes were determined from the pressure dependence of the rate constants of
P H Lin et al.
Toxicology and applied pharmacology, 145(2), 399-408 (1997-08-01)
The dosimetry of chlorinated quinones arising from metabolism of pentachlorophenol (PCP), in the livers of male Sprague-Dawley rats and B6C3F1 mice was investigated via measurements of cysteinyl protein adducts and estimates of the second-order reaction rate constants between the quinones
B Chakraborty et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 57(2), 223-229 (2001-02-24)
The equilibrium constants, enthalpies and entropies of formation of molecular electron donor-acceptor (EDA) complexes of o-chloranil with a series of aromatic hydrocarbons have been determined spectrophotometrically. Spectroscopic and thermodynamic aspects of these complexes have been analysed.
Hamid R Zare et al.
The journal of physical chemistry. B, 113(23), 8080-8085 (2009-05-21)
The electrochemical reduction of o-chloranil (OCA) in aqueous solution has been studied experimentally and theoretically. The effects of temperature and pH on various thermodynamic parameters were studied by means of cyclic voltammetry. The pH dependence of the redox activity of
Dalim Kumar Roy et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 61(9), 2017-2022 (2005-05-25)
Cloxacillin sodium has been shown to form charge transfer (CT) complexes of 1:1 stoichiometry with a number of electron acceptors in 50% (v/v) aqueous ethanol medium. From the trends in the CT absorption bands, the vertical ionization potential of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service