Skip to Content
Merck
All Photos(1)

Documents

T37001

Sigma-Aldrich

p-Toluic hydrazide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CONHNH2
CAS Number:
Molecular Weight:
150.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

116-118 °C (lit.)

SMILES string

Cc1ccc(cc1)C(=O)NN

InChI

1S/C8H10N2O/c1-6-2-4-7(5-3-6)8(11)10-9/h2-5H,9H2,1H3,(H,10,11)

InChI key

MFFVZXOPRXMVET-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jeya Shree Ganesan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 219, 33-43 (2019-04-29)
A simple julolidine based chemosensor (JT) was designed and synthesized by single condensation step. JT displayed excellent selectivity and sensitivity with on-off responses towards Zn2+ and Cu2+ over other biologically relevant metal ions in aqueous media. Upon addition of Zn2+

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service