SYNA381420
Synthware™ 90 degree vacuum adapter with PTFE stopcock
male joint: ST/NS 14/20
Synonym(s):
Synthware A381420, glass adapter with stopcock
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
male joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A381420
Related Categories
General description
With 8 mm or 10 mm O.D. hose connection and 2 mm PTFE stopcock.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)