Skip to Content
Merck
All Photos(1)

Key Documents

Q51545

Sigma-Aldrich

Urea-15N2

2 atom % 15N, 99% (CP)

Synonym(s):

15N Labeled urea, Carbamide-15N2, Carbonyldiamide-15N2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H215NCO15NH2
CAS Number:
Molecular Weight:
62.04
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

2 atom % 15N

Assay

99% (CP)

mol wt

60.09 by atom % calculation (2 atom % 15N)

mp

132-135 °C (lit.)

application(s)

agriculture

mass shift

M+2

SMILES string

[15NH2]C([15NH2])=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i2+1,3+1

InChI key

XSQUKJJJFZCRTK-SUEIGJEOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service