Skip to Content
Merck
All Photos(3)

Documents

P73005

Sigma-Aldrich

Pyromellitic diimide

97%

Synonym(s):

Benzene-1,2,4,5-tetracarboxylic diimide, Reillex® 402

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H4N2O4
CAS Number:
Molecular Weight:
216.15
Beilstein:
214780
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

>320 °C (lit.)

SMILES string

O=C1NC(=O)c2cc3C(=O)NC(=O)c3cc12

InChI

1S/C10H4N2O4/c13-7-3-1-4-6(10(16)12-8(4)14)2-5(3)9(15)11-7/h1-2H,(H,11,13,15)(H,12,14,16)

InChI key

UGQZLDXDWSPAOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Reillex is a registered trademark of Vertellus Specialties, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor V Rubtsov et al.
Journal of the American Chemical Society, 126(9), 2684-2685 (2004-03-05)
We demonstrate for the first time the utility of time-resolved visible pump/mid-infrared (IR) probe spectroscopy to interrogate directly, and provide unique infomation regarding, conformationally dependent photoinduced ET dynamics and the subsequent structural evolution of the resulting charge-separated (CS) state. Exemplary
Shin-ichiro Kato et al.
The Journal of organic chemistry, 71(13), 4723-4733 (2006-06-17)
This paper reports the electroscopic and electrochemical properties of [2 + 2] pyromellitic diimide-based cyclophane 1 as well as acyclic N,N'-bis(2-methoxybenzyl)pyromellitic diimide 2 and the clathrate compounds formed by 1. Compound 1 was synthesized by direct cyclocondensation. Its structure was
Bijitha Balan et al.
Organic letters, 9(14), 2709-2712 (2007-06-07)
Complex formation of pyromellitic diimide derivatives with beta-cyclodextrin and anthracene-appended beta-cyclodextrin was studied with use of induced circular dichroism and 1H NMR spectroscopies. It is revealed that pyromellitic diimides form rim-binding type complexes with beta-CD and in these complexes the
S A Martin et al.
Journal of animal science, 60(2), 544-550 (1985-02-01)
The effects of monensin, 2-bromoethanesulfonic acid (2-BES) and pyromellitic diimide (diimide) on gas and volatile fatty acid (VFA) production by the rumen microbiota were compared in mixed culture. Oat hay, a hay-concentrate mixture (48% hay, 43% concentrate) and a soluble
Rocío García-Bueno et al.
Dalton transactions (Cambridge, England : 2003), 39(24), 5728-5736 (2010-05-25)
The hydroxo-complexes [Ni(2)(mcN(3))(2)(mu-OH)](2)(PF(6))(2)] [mcN(3) = 2,4,4-trimethyl-1,5,9-triazacyclododec-1-ene (Me(3)-mcN(3)) or 2,4,4,9-tetramethyl-1,5,9-triazacyclododec-1-ene (Me(4)-mcN(3))] react with imides (Him) in 1 : 2 (succinimide, glutarimide, saccharine and 1,8-naphthalimide) or 1 : 1 (pyromellitic diimide) molar ratio, leading to the formation of the imidate complexes: [Ni(mcN(3))(H(2)O)(Im)](+)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service