Skip to Content
Merck
All Photos(5)

Documents

P33402

Sigma-Aldrich

2-Phenylpyridine

greener alternative

98%

Synonym(s):

α-Phenylpyridine, o-Phenylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9N
CAS Number:
Molecular Weight:
155.20
Beilstein:
110445
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

greener alternative product score

old score: 2
new score: 1
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.623 (lit.)

bp

268-270 °C (lit.)

density

1.086 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

c1ccc(cc1)-c2ccccn2

InChI

1S/C11H9N/c1-2-6-10(7-3-1)11-8-4-5-9-12-11/h1-9H

InChI key

VQGHOUODWALEFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Atom Economy”, “Design for Energy Efficiency” and “Use of Renewable Feedstock”. Click here to view its DOZN scorecard.

Application

Versatile synthetic intermediate.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huazhou Wu et al.
Dalton transactions (Cambridge, England : 2003), 40(9), 1969-1976 (2011-02-02)
Iridium(III) complexes with intense phosphorescence in solution have been widely applied in organic light-emitting diodes, chemosensors and bioimaging. However, little attention has been paid to iridium(III) complexes showing weak phosphorescence in solution and enhanced phosphorescence emission in the solid state
Palladium-catalyzed methylation of aryl C-H bond by using peroxides.
Yuhua Zhang et al.
Journal of the American Chemical Society, 130(10), 2900-2901 (2008-02-14)
Joeri Kuil et al.
Chembiochem : a European journal of chemical biology, 12(12), 1897-1903 (2011-07-09)
The chemokine receptor 4 (CXCR4) is over-expressed in 23 types of cancer in which it plays a role in, among others, the metastatic spread. For this reason it is a potential biomarker for the field of diagnostic oncology. The antagonistic
Michael T Vagnini et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(39), 15651-15656 (2012-05-16)
Photodriving the activity of water-oxidation catalysts is a critical step toward generating fuel from sunlight. The design of a system with optimal energetics and kinetics requires a mechanistic understanding of the single-electron transfer events in catalyst activation. To this end
T Okada et al.
Journal of medicinal chemistry, 44(26), 4661-4667 (2001-12-14)
Mouse sarcoma 180 cell with a 25-fold higher cisplatin (CDDP) resistance, termed S-180cisR, is newly established. S-180cisR cells grow quite slowly in the presence of CDDP with high concentration. This may show that S-180cisR cells modulate the cell cycle to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service