Skip to Content
Merck
All Photos(2)

Key Documents

P24152

Sigma-Aldrich

N-Phenylethylenediamine

98%

Synonym(s):

N-(2-Aminoethyl)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCH2CH2NH2
CAS Number:
Molecular Weight:
136.19
Beilstein:
508205
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.587 (lit.)

bp

262-264 °C (lit.)

density

1.041 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

NCCNc1ccccc1

InChI

1S/C8H12N2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,10H,6-7,9H2

InChI key

OCIDXARMXNJACB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Fan et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(1), 1801368-1801368 (2019-01-16)
Disperse reactive dyes with appropriate chemical structure are key for the coloration of natural fibers in the water-free environmentally friendly medium of supercritical carbon dioxide with various advantages. The objective of this work is to design and synthesize a novel
Babli Kumari et al.
Dalton transactions (Cambridge, England : 2003), 47(39), 14008-14016 (2018-09-20)
Structurally characterised two polymeric complexes of Cu(ii) (3) and Mn(iii) (4) of the ligand L (2-(((2-(phenylamino)ethyl)imino)methyl)phenol) have been explored for catecholase-like activity, fluorescence recognition of histidine and catalytic activity towards coupling of aryl iodides with benzamide, leading to N-arylbenzamides. CuO
Sakthi Raje et al.
Chemistry, an Asian journal, 13(11), 1458-1466 (2018-04-01)
Koneramines (LR OR', R=Ph or Ts; R'=Me, iPr) and their complexes were found to emerge from the system of pyridine-2-carboxaldehyde and N-phenyl/tosylethylenediamine when a primary or secondary alcohol was used as solvent. Imidazolidinylpyridines (LR , R=Ph or Ts) became major
Leslie S Hamachi et al.
Chemical science, 10(26), 6539-6552 (2019-08-02)
We report a method to control the composition and microstructure of CdSe1-x S x
Abhijit Ghosh et al.
Dalton transactions (Cambridge, England : 2003), 44(26), 11797-11804 (2015-06-09)
A very simple molecule derived from salicylaldehyde and N-phenyl ethylenediamine (L1) functions as dual-mode ratiometric fluorescence "turn on" sensors for Zn(2+) and Al(3+) at two different wavelengths. The sensing is based on the combined effect of inhibition of excited-state intra-molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service