Skip to Content
Merck
All Photos(1)

Key Documents

N27802

Sigma-Aldrich

4-Nitro-3-(trifluoromethyl)phenol

99% (GC)

Synonym(s):

Lamprecide, TFM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(CF3)OH
CAS Number:
Molecular Weight:
207.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99% (GC)

form

powder

bp

135-138 °C/0.01 mmHg (lit.)

mp

76-79 °C (lit.)

SMILES string

Oc1ccc(c(c1)C(F)(F)F)[N+]([O-])=O

InChI

1S/C7H4F3NO3/c8-7(9,10)5-3-4(12)1-2-6(5)11(13)14/h1-3,12H

InChI key

ZEFMBAFMCSYJOO-UHFFFAOYSA-N

Gene Information

human ... CA2(760)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oana Birceanu et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 153(3), 342-349 (2010-12-22)
The toxicity of 3-trifluoromethyl-4-nitrophenol (TFM) appears to be due to a mismatch between ATP supply and demand in lamprey, depleting glycogen stores and starving the nervous system of ATP. The cause of this TFM-induced ATP deficit is unclear. One possibility
Hongwei Hu et al.
Sensors (Basel, Switzerland), 18(6) (2018-06-08)
The ultrasonic phased array total focusing method (TFM) has the advantages of full-range dynamic focusing and high imaging resolution, but the problem of long imaging time limits its practically industrial applications. To reduce the imaging calculation demand of TFM, the
L V Magnussen et al.
Andrology, 5(5), 946-953 (2017-09-16)
The purpose of the study was to evaluate whether testosterone replacement therapy improves muscle mechanical and physical function in addition to increasing lean leg mass and total lean body mass in aging men with type 2 diabetes and lowered bio-available
Kathryn A Rosowski et al.
Scientific reports, 8(1), 17967-17967 (2018-12-21)
Cells respond to the mechanics of their environment. Mechanical cues include extracellular matrix (ECM) stiffness and deformation, which are primarily sensed through integrin-mediated adhesions. We investigated the impact of ECM deformation on cellular forces, measuring the time-evolution of traction forces
Luís B Sardinha et al.
Pediatric obesity, 12(5), 373-379 (2016-06-04)
The amount of time children spend in sedentary behaviours may have adverse health effects. To examine the substitution effects of displacing a fixed duration of sedentary time with physical activity (PA) on children's body composition. We included 386 children (197

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service