Skip to Content
Merck
All Photos(2)

Key Documents

N22851

Sigma-Aldrich

1-Nitropropane

≥98.5%

Synonym(s):

1-Nitropropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2NO2
CAS Number:
Molecular Weight:
89.09
Beilstein:
506236
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.1 (vs air)

vapor pressure

7.5 mmHg ( 20 °C)

Assay

≥98.5%

form

liquid

autoignition temp.

788 °F

IVD

for in vitro diagnostic use

refractive index

n20/D 1.401 (lit.)

bp

131-132 °C (lit.)

density

0.998 g/mL at 25 °C (lit.)

SMILES string

CCC[N+]([O-])=O

InChI

1S/C3H7NO2/c1-2-3-4(5)6/h2-3H2,1H3

InChI key

JSZOAYXJRCEYSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E George et al.
Carcinogenesis, 10(12), 2329-2334 (1989-12-01)
2-Nitropropane (2-NP) is a rat liver carcinogen, whilst the 1-isomer is non-carcinogenic in rodents. Although DNA repair tests in the rat liver discriminated clearly between the carcinogenic and the non-carcinogenic isomer, uniformly negative results have been published for the mouse
E S Fiala et al.
Carcinogenesis, 8(12), 1947-1949 (1987-12-01)
1-Nitropropane (1-NP), 2-nitropropane (2-NP), 1-azoxypropane (1-AP) and 2-azoxypropane (2-AP), were assayed for carcinogenicity by gavage in male Sprague-Dawley rats. 2-NP was given at 1 mmol/kg three times per week for 16 weeks. 1-NP (1 mmol/kg), 1-AP (0.1 mmol/kg), 2-AP (0.1
Inhalation exposure of rats to vapors of 1-nitropropane at 100 ppm.
T B Griffin et al.
Ecotoxicology and environmental safety, 6(3), 268-282 (1982-06-01)
U Kliesch et al.
Mutation research, 192(3), 181-184 (1987-11-01)
Micronucleus tests were carried out in bone marrow of mice treated with 1-nitropropane, 2-nitropropane and cisplatin. For 1-nitropropane and 2-nitropropane the results were negative. With cisplatin a dose-dependent increase in the number of polychromatic erythrocytes with micronuclei was observed. The
Anna Fryszkowska et al.
The FEBS journal, 279(22), 4160-4171 (2012-09-18)
Enzymes are natural catalysts, controlling reactions with typically high stereospecificity and enantiospecificity in substrate selection and/or product formation. This makes them useful in the synthesis of industrially relevant compounds, particularly where highly enantiopure products are required. The flavoprotein pentaerythritol tetranitrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service