Skip to Content
Merck
All Photos(2)

Key Documents

N1658

Sigma-Aldrich

1,8-Naphthalimide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7NO2
CAS Number:
Molecular Weight:
197.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

299-300 °C (lit.)

SMILES string

O=C1NC(=O)c2cccc3cccc1c23

InChI

1S/C12H7NO2/c14-11-8-5-1-3-7-4-2-6-9(10(7)8)12(15)13-11/h1-6H,(H,13,14,15)

InChI key

XJHABGPPCLHLLV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiaojiao Liu et al.
Colloids and surfaces. B, Biointerfaces, 170, 656-662 (2018-07-10)
Highly ordered self-assembly of nanoparticles (NPs) in a large scale promises attractive potential in optical modulation of the NPs for illuminating, imaging and sensing applications. In this work, a type of multi-lamellar nanocomposite membranes composed of phospholipid multilayers and Si
Zhen Wu et al.
Small (Weinheim an der Bergstrasse, Germany), 14(40), e1802524-e1802524 (2018-10-12)
Organic luminescent materials with the ability to reversibly switch the luminescence when subjected to external stimuli have attracted considerable interest in recent years. However, the examples of luminescent materials that exhibit multiresponsive properties are rarely reported. In this work, a
Yan Guo et al.
Inorganic chemistry, 58(19), 13150-13160 (2019-09-21)
Mitochondrial DNA (mtDNA) is an attractive cellular target for anticancer agents in addition to nuclear DNA (nDNA). The cationic platinum(II) complex cis-[Pt(NP)(NH3)2Cl]NO3 (PtNP, NP = N-(2-ethylpyridine)-1,8-naphthalimide) bearing the DNA-intercalating moiety NP was designed. The structure of PtNP was fully characterized
Xiaoyuan Ji et al.
Nanoscale, 10(30), 14455-14463 (2018-07-20)
The development of biocompatible and fluorescent gene carriers is of particular importance in the gene-delivery field. Taking advantage of the unique optical properties (e.g., strong and robust fluorescence) of silicon nanoparticles (SiNPs), as well as the excellent biocompatibility of silicon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service