Skip to Content
Merck
All Photos(1)

Key Documents

M89803

Sigma-Aldrich

Mucochloric acid

99%

Synonym(s):

2,3-Dichloromalealdehydic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHCCCl=CClCOOH
CAS Number:
Molecular Weight:
168.96
Beilstein:
1705641
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

crystals

mp

125-128 °C (lit.)

SMILES string

OC(=O)\C(Cl)=C(/Cl)C=O

InChI

1S/C4H2Cl2O3/c5-2(1-7)3(6)4(8)9/h1H,(H,8,9)/b3-2+

InChI key

LUMLZKVIXLWTCI-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

366.8 °F

Flash Point(C)

186 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I L Steffensen et al.
Pharmacology & toxicology, 85(2), 56-64 (1999-09-17)
Epidemiological studies indicate an association between exposure to chlorinated drinking water and risk of intestinal cancer. In order to study this experimentally, we have examined the effects of 3,4-dichloro-5-hydroxy-2[5H]-furanone (mucochloric acid, MCA) and 3-chloro-4-(dichloromethyl)-5-hydroxy-2[5H]-furanone (MX), mutagenic and genotoxic compounds in
R T LaLonde et al.
Chemical research in toxicology, 5(5), 618-624 (1992-09-01)
The Salmonella typhimurium (TA100) mutagenic compound, mucochloric acid [3,4-dichloro-5-hydroxy-2(5H)-furanone (MCA)], was inactivated by in vitro N-acetylcysteine (NAC). The reaction of MCA with NAC at pH7 was second order and gave products 4, 5, and 6a that resulted from the displacement
D Asplund et al.
Chemical research in toxicology, 8(6), 841-846 (1995-09-01)
The bacterial mutagen mucochloric acid was reacted with adenosine in aqueous solutions at 37 degrees C. In the HPLC chromatograms of the reaction mixtures two peaks of unidentified products were observed at longer retention times than the previously characterized "etheno"
B Isomaa et al.
Toxicology, 100(1-3), 69-77 (1995-06-26)
The cytotoxicity of the, in Salmonella, potent mutagenic compound, 3-chloro-4-(dichloromethyl)-5-hydroxy-2(5H)-furanone (MX) and its structural analogue 3,4-dichloro-5-hydroxy-2[5H]-furanone (mucochloric acid, MCA), was studied in freshly isolated rainbow trout hepatocytes and gill epithelial cells by determining 86Rb-leakage and decrease in fluorescence intensity in
R T LaLonde et al.
Chemical research in toxicology, 7(4), 482-486 (1994-07-01)
The inactivation of the drinking water mutagen mucochloric acid (MCA) by reduced glutathione (GSH) was linked to the formation of an MCA-GSH conjugate, a nonmutagen in the Salmonella typhimurium (TA100) plate incorporation assay. Anaerobic formation of MCA-GSH is found now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service