M89617
Mucic acid
97%
Synonym(s):
Galactaric acid, MTPA, Saccharolactic acid, Tetrahydroxyadipic acid, Tetrahydroxyhexanedioic acid
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
HOOC(CHOH)4COOH
CAS Number:
Molecular Weight:
210.14
Beilstein:
1728117
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
powder
mp
220-225 °C (dec.) (lit.)
SMILES string
O[C@@H]([C@@H](O)[C@H](O)C(O)=O)[C@@H](O)C(O)=O
InChI
1S/C6H10O8/c7-1(3(9)5(11)12)2(8)4(10)6(13)14/h1-4,7-10H,(H,11,12)(H,13,14)/t1-,2+,3+,4-
InChI key
DSLZVSRJTYRBFB-DUHBMQHGSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Mucic acid or galactaric acid can be used as a precursor for the synthesis of:
It can be also utilized in the surface modification of monodisperse water-soluble magnetic nanoparticles.
- 2,3,4,5-tetra-O-acetylgalactaric acid (AGA) which is used as a monomer unit along with adipic acid for the synthesis of copolyanhydrides.
- Muconic acid and adipic acid.
It can be also utilized in the surface modification of monodisperse water-soluble magnetic nanoparticles.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Highly Efficient Chemical Process To Convert Mucic Acid into Adipic Acid and DFT Studies of the Mechanism of the Rhenium-Catalyzed Deoxydehydration
Li X, et al.
Angewandte Chemie (International ed. in English), 53(16), 4200-4204 (2014)
Functionalization of monodisperse magnetic nanoparticles
Lattuada M and Hatton T A
Langmuir, 23(4), 2158-2168 (2007)
C Lavilla et al.
Biomacromolecules, 12(7), 2642-2652 (2011-05-14)
The dimethyl ester of 2,3:4,5-di-O-methylene-galactaric acid (Galx) was made to react in the melt with 1,n-alkanediols HO(CH(2))(n)OH containing even numbers of methylenes (n from 6 to 12) to produce linear polycyclic polyesters. Two sets of poly(alkylene 2,3:4,5-di-O-methylene-galactarate) polyesters (PE-nGalx) with
Tanja Spoettl et al.
International journal of colorectal disease, 22(3), 303-312 (2006-05-23)
Nicotine is of therapeutic value in ulcerative colitis, but its administration is connected with adverse events. Nicotine derivatives are currently being tested to maintain the therapeutic effects and minimize adverse events. TC-2403-12 is a (E)-metanicotine hemigalactarate. The aim of this
W Tian et al.
Carbohydrate research, 324(1), 45-52 (2000-03-21)
The FT-IR spectra of galactaric acid and its K+, NH4+, Ca2+, Ba2+, and La3+ salts have been recorded and interpreted. Spectroscopic evidence shows that the dimeric carboxylic groups of the free acid are dissociated upon formation of the salt, and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service