Skip to Content
Merck
All Photos(1)

Documents

M3507

Sigma-Aldrich

2-Mercaptobenzoxazole

95%

Synonym(s):

2-Benzoxazolethiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NOS
CAS Number:
Molecular Weight:
151.19
Beilstein:
119483
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

mp

192-195 °C (lit.)

SMILES string

Sc1nc2ccccc2o1

InChI

1S/C7H5NOS/c10-7-8-5-3-1-2-4-6(5)9-7/h1-4H,(H,8,10)

InChI key

FLFWJIBUZQARMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohui Gao et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 3(12), 1600126-1600126 (2016-12-17)
Organic ligand-protected metal nanoclusters have attracted extensively attention owing to their atomically precise composition, determined atom-packing structure and the fascinating properties and promising applications. To date, most research has been focused on thiol-stabilized gold and silver nanoclusters and their single
D Zimmermann et al.
The Journal of membrane biology, 221(2), 107-121 (2008-01-17)
Interactions of structurally dissimilar anionic compounds with the plasma membrane of HEK293 cells were analyzed by patch clamp and electrorotation. The combined approach provides complementary information on the lipophilicity, preferential affinity of the anions to the inner/outer membrane leaflet, adsorption
Vera Klimesová et al.
European journal of medicinal chemistry, 44(5), 2286-2293 (2008-08-13)
A set of 2-benzylsulfanyl derivatives of benzoxazole was synthesized and evaluated for their in vitro antimycobacterial activity against Mycobacterium tuberculosis, non-tuberculous mycobacteria and multidrug-resistant M. tuberculosis. The activities were expressed as the minimum inhibitory concentration (MIC) in mmol/L. The substances
G Peng et al.
British journal of cancer, 103(4), 542-551 (2010-07-22)
Tumour growth is accompanied by gene and/or protein changes that may lead to peroxidation of the cell membrane species and, hence, to the emission of volatile organic compounds (VOCs). In this study, we investigated the ability of a nanosensor array
Tesfalem Geremariam Welearegay et al.
Nanotechnology, 29(6), 065603-065603 (2017-12-06)
Here, we report for the first time the fabrication of ligand-functionalised ultrapure monodispersed metal nanoparticles (Au, Cu, and Pt) from their pure metal precursors using the advanced gas deposition technique. The experimental conditions during nanoparticle formation were adjusted in order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service