Skip to Content
Merck
All Photos(1)

Documents

L3501

Sigma-Aldrich

2,3-Lutidine

99%

Synonym(s):

2,3-Dimethylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.508 (lit.)

bp

162-163 °C (lit.)

mp

−15 °C (lit.)

density

0.945 g/mL at 25 °C (lit.)

SMILES string

Cc1cccnc1C

InChI

1S/C7H9N/c1-6-4-3-5-8-7(6)2/h3-5H,1-2H3

InChI key

HPYNZHMRTTWQTB-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuka Fujito et al.
Journal of chromatography. A, 1626, 461363-461363 (2020-08-17)
Analytical techniques to determine volatile compounds such as flavor, aroma, and fragrances are in high demand due to their wide range of applications in industry, the chemical properties of them are very diverse. Supercritical fluid chromatography (SFC) is capable of
Zhiming Sun et al.
Journal of colloid and interface science, 511, 268-276 (2017-10-17)
To overcome rapid electron-hole recombination and low adsorption capacity of single g-C
Lucia Cancelada et al.
Environmental science & technology, 53(13), 7866-7876 (2019-06-01)
This study characterized emissions from IQOS, a heated tobacco product promoted as a less harmful alternative to cigarettes. Consumable tobacco plugs were analyzed by headspace GC/MS to assess the influence of heating temperature on the emission profile. Yields of major
Tong Li et al.
Colloids and surfaces. B, Biointerfaces, 191, 111001-111001 (2020-04-24)
Metal organic frameworks (MOFs) served as peroxidase-like artificial enzymes have been recently adopted for wide applications including therapy, pollution degradation, biosensing and so on. However, most of MOFs mimicking peroxidase cannot perform with the utmost efficiency under certain biological circumstances
Xiaoyong Luo et al.
Pesticide biochemistry and physiology, 143, 258-264 (2017-12-01)
In order to understand the action mechanism of fluazifop-P-butyl (FB) in bristly starbur (Acanthospermum hispidum D.C.), a susceptible plant, the role of active oxygen species (ROS) in herbicide-induced cell death in shoots was investigated. FB-induced phytotoxicity was not reduced by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service