Regarding KITALYSIS-CN-1KT KitAlysis™ High-Throughput Buchwald-Hartwig Amination Reaction Screening Kit-Pack of 4, there are links to specific usage instructions posted on this page.
https://www.sigmaaldrich.com/product/aldrich/kitalysiscn#product-documentation
These describe reactions carried out at various temperatures (depending on the substrate used), e.g. 60 - 80 degrees C, controlled by the use of a heating bath.
KITALYSIS-CN
KitAlysis™ High-Throughput Buchwald-Hartwig Amination Reaction Screening Kit-Pack of 4
Synonym(s):
Amination Screening Kit, Buchwald-Hartwig Amination
About This Item
Recommended Products
reaction suitability
reaction type: Buchwald-Hartwig Amination
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reagent type: catalyst
core: palladium
storage temp.
room temp
General description
Application
Two Solvents, Weak Base (Cs2CO3)
Download the experimental excel file here
Two Solvents, Strong Base (NaOt-Bu)
Download the experimental excel file here
One Solvent (Dioxane), Two Bases
Download the experimental excel file here
One Solvent (DMAc), Two Bases
Download the experimental excel file here
Each kit contains 4 screening sets that contain the following:
Each customizable screening kit contains
- 12 X 2 pre-weighed catalysts (1 μmol) in individual microvials with magnetic stir bars
- 2 bases (NaOt-Bu and Cs2CO3)
- 2 degassed anhydrous solvents (dioxane and DMAc) in ampules
- 2 substrate reaction vials with stir bars
- 1 internal standard
- 4 reaction block replacement films
Best when used with KitAlysis Benchtop Inertion Box and 24-well Reaction Block (sold separately).
Features and Benefits
- Designed and tested by synthetic chemists
- Easily run 24-microscale reaction in parallel
- 10 μmol reactions with 1 μmol catalyst, 3 equiv. base, 0.1 M concentration
- Customizable Excel Template for quick calculations and easy incorporation into electronic laboratory notebooks
Legal Information
Kit Components Only
- SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules 1 EA
Kit Components Also Available Separately
- 900634N,N-Dimethylacetamide, anhydrous, ZerO2®, 99.8% 8 mLSDS
- 9006401,4-Dioxane, anhydrous, ZerO2®, 99.8% 8 mLSDS
- 702706Sodium tert-butoxide solution, 2 M in THF, ZerO2® 4 mLSDS
- 747130Josiphos SL-J009-1 Pd G3 5 µmolSDS
- 753009SPhos Pd G2 5 µmolSDS
- 753246RuPhos Pd G2 5 µmolSDS
- 756482P(t-Bu)3 Pd G2 5 µmolSDS
recommended
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Repr. 1B - Self-heat. 1 - Skin Corr. 1B - STOT RE 2 Oral - STOT SE 3
Target Organs
Central nervous system, Kidney,Adrenal gland,Testes, Respiratory system
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
Flash Point(F)
-4.0 °F - closed cup
Flash Point(C)
-20 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Protocol for the KitAlysis™ Buchwald-Hartwig Amination Reaction Screening Kit
Protocol for the KitAlysis™ Buchwald-Hartwig Amination Reaction Screening Kit
Protocol for the KitAlysis™ Buchwald-Hartwig Amination Reaction Screening Kit
Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.
Related Content
Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.
Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.
Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.
Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.
-
Good morning Iwould have a question regarding the KitAlysis™ High-Throughput Buchwald-Hartwig Amination Reaction Screening Kit-Pack of 4. Is It possible to work at various temperatures or it's only thinked for the RT?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service