Skip to Content
Merck
All Photos(4)

Key Documents

K6000

Sigma-Aldrich

α-Keto-γ-(methylthio)butyric acid sodium salt

≥97%

Synonym(s):

4-Methylthio-2-oxobutanoic acid sodium salt, 4-Methylthio-2-oxobutyric acid sodium salt, KMBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H7O3SNa
CAS Number:
Molecular Weight:
170.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

storage temp.

2-8°C

SMILES string

[Na].CSCCC(=O)C(O)=O

InChI

1S/C5H8O3S.Na/c1-9-3-2-4(6)5(7)8;/h2-3H2,1H3,(H,7,8);/q;+1/p-1

InChI key

IFSCKRWNXKWTLR-UHFFFAOYSA-M

General description

α-Keto-γ-methylthiobutyric acid (KMTB) is a keto derivative of L-methionine that shows considerable potential as a substitute for L-methionine. The sodium salt of KMTB can be employed in nucleophilic substitution reactions. Additionally, it acts as a building block for the synthesis of amino acids, peptides, and other complex organic compounds.

Application


  • Measuring antioxidant capacity using the ORAC and TOSC assays.: This study details the use of α-Keto-γ-(methylthio)butyric acid sodium salt in assessing antioxidant capacity through ORAC and TOSC assays, emphasizing its utility in metabolic and chemical synthesis research (Garrett AR et al., 2010).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-step biosynthesis of $\alpha$-keto-$\gamma$-methylthiobutyric acid from L-methionine by an Escherichia coli whole-cell biocatalyst expressing an engineered L-amino acid deaminase from Proteus vulgaris
Hossain, Gazi Sakir and Li, et al.
PLoS ONE, 9, e114291-e114291 (2014)
Simona Daniele et al.
Frontiers in aging neuroscience, 10, 17-17 (2018-02-15)
The loss of protein homeostasis that has been associated with aging leads to altered levels and conformational instability of proteins, which tend to form toxic aggregates. In particular, brain aging presents characteristic patterns of misfolded oligomers, primarily constituted of β-amyloid
Charlotte Sandersen et al.
Veterinary anaesthesia and analgesia, 38(2), 83-93 (2011-02-10)
To investigate the effect of acepromazine (ACP) on reactive oxygen species (ROS) production by stimulated equine neutrophils. Ex vivo biochemical experiments. Isolated neutrophils from healthy untreated horses. Neutrophils were incubated with ACP at concentrations of 10(-4), 10(-5) or 10(-6) M
F Regoli et al.
Toxicology and applied pharmacology, 156(2), 96-105 (1999-04-13)
We have extended the application of our previously reported total oxidant scavenging capacity (TOSC) assay (Winston et al., Free Radical Biol. Med. 24, 480-493, 1998) to permit facile quantification of the absorbance capacity of antioxidants toward three potent oxidants, i.e.
Gary W Winston et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 139(4), 281-288 (2005-02-03)
During embryogenesis in grass shrimp the capacity to scavenge oxyradicals increased as measured by the Total Oxyradical Scavenging Capacity (TOSC) assay. The increase in TOSC during embryogenesis was associated with increasing concentrations of a number of antioxidants, including coenzyme Q

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service