Skip to Content
Merck
All Photos(1)

Key Documents

I2401

Sigma-Aldrich

Indazole

98%

Synonym(s):

1,2-Benzodiazole, 1,2-Benzopyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2
CAS Number:
Molecular Weight:
118.14
Beilstein:
109676
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

bp

270 °C/743 mmHg (lit.)

mp

145-148 °C (lit.)

SMILES string

c1ccc2[nH]ncc2c1

InChI

1S/C7H6N2/c1-2-4-7-6(3-1)5-8-9-7/h1-5H,(H,8,9)

InChI key

BAXOFTOLAUCFNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Ling Wang et al.
Journal of fluorescence, 30(5), 1105-1112 (2020-07-09)
The widespread occurrence of sulfonamides (SAs) in the environment water has rasied great concerns about their potential to antibiotics resistance. In this study, the fluorescence behaviors of sulfamethoxazole (SMZ) representing certain properties of the SAs mixed with three different kinds
Gabriel E Büchel et al.
Inorganic chemistry, 52(11), 6273-6285 (2013-05-11)
Ruthenium nitrosyl complexes of the general formulas (cation)(+)[cis-RuCl4(NO)(Hazole)](-), where (cation)(+) = (H2ind)(+), Hazole = 1H-indazole (Hind) (1c), (cation)(+) = (H2pz)(+), Hazole = 1H-pyrazole (Hpz) (2c), (cation)(+) = (H2bzim)(+), Hazole = 1H-benzimidazole (Hbzim) (3c), (cation)(+) = (H2im)(+), Hazole = 1H-imidazole (Him)
Anatolie Gavriluta et al.
Inorganic chemistry, 52(11), 6260-6272 (2013-05-17)
Synthesis and X-ray diffraction structures of cis and trans isomers of ruthenium and osmium metal complexes of general formulas (nBu4N)[cis-MCl4(NO)(Hind)], where M = Ru (1) and Os (3), and (nBu4N)[trans-MCl4(NO)(Hind)], where M = Ru (2) and Os (4) and Hind
Kornelia Czaja et al.
International journal of molecular sciences, 21(13) (2020-07-11)
Vascular endothelial growth factor receptor 2 (VEGFR2) is a key receptor in the angiogenesis process. The VEGFR2 expression is upregulated in many cancers so this receptor is an important target for anticancer agents. In the present paper, we analyse interactions
Rolf Spirig et al.
PloS one, 5(6), e0010983-e0010983 (2010-06-12)
Dendritic cells (DC) are professional antigen presenting cells that represent an important link between innate and adaptive immunity. Danger signals such as toll-like receptor (TLR) agonists induce maturation of DC leading to a T-cell mediated adaptive immune response. In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service