Skip to Content
Merck
All Photos(2)

Key Documents

H6800

Sigma-Aldrich

1-Hexadecanol

95%

Synonym(s):

Cetyl alcohol, Palmityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
Beilstein:
1748475
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
95%
bp:
179-181 °C/10 mmHg (lit.)
vapor pressure:
<0.01 mmHg ( 43 °C)
Pricing and availability is not currently available.

vapor density

8.34 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 43 °C)

Assay

95%

form

solid

autoignition temp.

483 °F

expl. lim.

8 %

bp

179-181 °C/10 mmHg (lit.)

mp

48-50 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCO

InChI

1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

BXWNKGSJHAJOGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • 1-Hexadecanol can be used as a reactant in the synthesis of self-assembled m-diethynylbenzene macrocycles (DBMs)[1], water-soluble pillar[6]arene for controllable drug release[2] and poly(ethylene glycol)-based polymer for intracellular delivery of anticancer drugs.[3]
  • It is used in building the template for sol-gel synthesis of mesoporous silicates.[4]
  • It can be employed in the synthesis of anionic gemini surfactants which also exhibit antibacterial and antifungal activities.[5]
  • High-chain fatty acid esters of 1-hexadecanol can be used as phase change materials (PCM) for thermal energy storage.[6]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization, surface properties and biological activity of some synthesized anionic surfactants.
Negm N A and Tawfik S M
Journal of Industrial and Engineering Chemistry, 20(6), 4463-4472 (2014)
Cellular uptake, intracellular trafficking, and antitumor efficacy of doxorubicin-loaded reduction-sensitive micelles.
Cui C, et al.
Biomaterials, 34(15), 3858-3869 (2013)
Aikaterini Papadaki et al.
Biomolecules, 10(1) (2020-01-16)
Oleogelation is an emerging technology to structure oils, which can be widely used to substitute saturated and trans fats. Extra virgin olive oil is widely recognized for its high nutritional value, but its utilization in oleogel production is currently limited.
m-Diethynylbenzene macrocycles: syntheses and self-association behavior in solution.
Tobe Y, et al.
Journal of the American Chemical Society, 124(19), 5350-5364 (2002)
Jennifer M MacLeod et al.
ACS nano, 11(9), 8901-8909 (2017-08-15)
Two-dimensional (2D) molecular self-assembly allows for the formation of well-defined supramolecular layers with tailored geometrical, compositional, and chemical properties. To date, random intermixing and entropic effects in these systems have largely been associated with crystalline disorder and glassy phases. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service