Skip to Content
Merck
All Photos(3)

Documents

H12552

Sigma-Aldrich

Hexatriacontane

98%

Synonym(s):

n-Hexatriacontane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)34CH3
CAS Number:
Molecular Weight:
506.97
Beilstein:
1712367
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

17.6 (vs air)

Quality Level

Assay

98%

bp

265 °C/1 mmHg (lit.)

mp

74-76 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C36H74/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-36H2,1-2H3

InChI key

YDLYQMBWCWFRAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anuar El Ouahabi et al.
Environmental science and pollution research international, 24(34), 26605-26614 (2017-09-29)
Analyses of trimethylsilyl-derivatized neutral lipids by gas chromatography coupled to mass spectrometry afford a wealth of information contained in the marine sediments. This information is useful for unraveling the processes associated with past climate changes such as those from glacial
Juan Jesús Molina-Rueda et al.
Plant physiology and biochemistry : PPB, 94, 181-190 (2015-06-27)
Transgenic hybrid poplars engineered to express ectopically the heterologous pine cytosolic GS1a display a number of significant pleiotropic phenotypes including enhanced growth, enhanced nitrogen use efficiency, and resistance to drought stress. The present study was undertaken in order to assess

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service