Skip to Content
Merck
All Photos(2)

Key Documents

GF97030403

Antimony

foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 95+%

Synonym(s):

Antimony, SB000120

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sb
CAS Number:
Molecular Weight:
121.76
MDL number:
UNSPSC Code:
12141704
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥95%

form

foil

manufacturer/tradename

Goodfellow 970-304-03

resistance

41.7 μΩ-cm

bp

1635 °C (lit.)

mp

630 °C (lit.)

density

6.69 g/mL at 25 °C (lit.)

SMILES string

[Sb]

InChI

1S/Sb

InChI key

WATWJIUSRGPENY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward R T Tiekink
Critical reviews in oncology/hematology, 42(3), 217-224 (2002-06-07)
The main group elements antimony and bismuth are used clinically, primarily for the treatment of Leishmaniasis (antimony) and ulcers (bismuth). Despite their medicinal efficacy, the exploration of the anti-cancer potential of antimony and bismuth compounds is not as well developed
Khatima Aït-Oudhia et al.
Parasitology research, 109(5), 1225-1232 (2011-07-30)
Leishmania is the causative agent of various forms of leishmaniasis, a significant cause of morbidity and mortality. The clinical manifestations of the disease range from self-healing cutaneous and mucocutaneous skin ulcers to a fatal visceral form named visceral leishmaniasis or
Ricardo Khouri et al.
The Journal of infectious diseases, 210(2), 306-310 (2014-02-11)
We show that increased plasma superoxide dismutase 1 (SOD1) levels are statistically significant predictors of the failure of pentavalent antimony treatment for cutaneous leishmaniasis caused by Leishmania braziliensis. In Leishmania amazonensis-infected patients, host SOD1 levels can be used to discriminate
Patricia Smichowski
Talanta, 75(1), 2-14 (2008-03-29)
This review summarizes and discusses the research carried out on the determination of antimony and its predominant chemical species in atmospheric aerosols. Environmental matrices such as airborne particulate matter, fly ash and volcanic ash present a number of complex analytical
Martin Wiener et al.
Journal of plastic, reconstructive & aesthetic surgery : JPRAS, 67(8), 1038-1044 (2014-06-15)
Recommended selective neck dissections for primary cutaneous tumours on the head and neck are based largely on clinical recurrence data acquired prior to the era of lymphatic mapping and sentinel lymph node (SLN) biopsy. This study aimed to examine lymphatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service