Skip to Content
Merck
All Photos(1)

Key Documents

GF77686521

Polyimide

(PI), film, thickness 0.025 mm, L 5 m, grade kapton hn

Synonym(s):

IM301200

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

form

film

manufacturer/tradename

Goodfellow 776-865-21

L

5 m

thickness

0.025 mm

General description

For updated SDS information please visit www.goodfellow.com
Polyimides are an important class of high-temperature engineering polymers consist of aromatic dianhydrides and diamine monomers. These polymers are well known for their high-temperature stability, low-temperature tolerance, mechanical strength, chemical resistance, radiation resistance, and excellent dielectric properties.

Application

Polyimide films (thickness 0.025 mm) can be used in capacitors, flexible printing circuits boards, insulation materials, and also in optoelectronic applications, such as transparent conductive film, transparent substrates for flexible display, and flexible solar cells. It is also used for the fabrication of flexible copper-clad laminates (FCCLs) for advanced microelectronic assembly.

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyimides
F.I.L./I.D.F., Milchwiss. (2017)
Polyimides
The Biological Effects of Glutamic Acid and Its Derivatives (2012)
A review on colorless and optically transparent polyimide films: Chemistry, process and engineering applications
Ni Hong-jiang, et al.
Journal of Industrial and Engineering Chemistry, 28, 16-27 (2015)
Study on charge transport mechanism and space charge characteristics of polyimide films
Zhou LR, et al.
IEEE Transactions on Dielectrics and Electrical Insulation, 16(4), 1143-1149 (2009)
A study of the composite insulation breakdown properties for wrapping cables in liquid nitrogen
Wei Bin, et al.
IEEE Transactions on Applied Superconductivity, 25(1), 1-6 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service