GF71666613
Tantalum
foil, 50x50mm, thickness 3.0mm, annealed, 99.9%
Synonym(s):
Tantalum, TA000580, Ta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
vapor pressure
<0.01 mmHg ( 537.2 °C)
Assay
99.9%
form
foil
autoignition temp.
572 °F
manufacturer/tradename
Goodfellow 716-666-13
resistivity
13.5 μΩ-cm, 20°C
size × thickness
50x50 mm × 3.0 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of hazardous materials, 225-226, 21-27 (2012-05-23)
In order to overcome the limitation of the application of nanoparticles, tantalum (oxy)nitrides nanotube arrays on a Ta foil were synthesized and introduced in vis (visible light)-Fenton-like system to enhance the degradation of atrazine. At first, the anodization of tantalum
Radiation research, 165(4), 452-459 (2006-04-04)
We determined effective cross sections for production of single-strand breaks (SSBs) in plasmid DNA [pGEM 3Zf(-)] by electrons of 10 eV and energies between 0.1 and 4.7 eV. After purification and lyophilization on a chemically clean tantalum foil, dry plasmid
Tantalum foil cuffs in peripheral nerve surgery.
Surgery, 23(3), 507-514 (1948-03-01)
Applied optics, 31(22), 4397-4404 (1992-08-01)
A simple four-element fused-silica lens is presented that has a focal length of 31.2 mm and a relative aperture of f/1 for use as a focusing lens for deep UV laser processing. The curvature of the lens is designed with
Tantalum foil pegs.
American journal of ophthalmology, 34(3), 444-444 (1951-03-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service