Skip to Content
Merck
All Photos(2)

Key Documents

GF51459217

Bismuth

foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 99.97%

Synonym(s):

Bismuth, BI000040

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99.97%

form

foil

manufacturer/tradename

Goodfellow 514-592-17

resistivity

129 μΩ-cm, 20°C

size × thickness

50x50 mm × 0.0025 mm

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge A R Salvador et al.
Molecules (Basel, Switzerland), 16(4), 2884-2913 (2011-04-06)
Steroid and terpene chemistry still have a great impact on medicinal chemistry. Therefore, the development of new reactions or "greener" processes in this field is a contemporaneous issue. In this review, the use of bismuth(III) salts, as "ecofriendly" reagents/catalysts, on
Hong Lu et al.
European journal of gastroenterology & hepatology, 25(10), 1134-1140 (2013-06-20)
Antimicrobial resistance has continued to undermine many popular anti-Helicobacter pylori therapies. Antibiotic resistance to commonly used anti-H. pylori drugs in China has increased markedly, making China an ideal site to identify regimens that remain effective despite widespread antimicrobial resistance. Bismuth
P Moayyedi et al.
Alimentary pharmacology & therapeutics, 17(10), 1215-1227 (2003-05-21)
Evidence for the effectiveness of antacids, histamine-2 receptor antagonists, bismuth salts, sucralfate and prokinetic therapy in non-ulcer dyspepsia is conflicting. To conduct a systematic review evaluating these therapies in non-ulcer dyspepsia. Electronic searches were performed using the Cochrane Controlled Trials
Andreia M C Rocha et al.
British journal of haematology, 168(3), 421-428 (2014-09-27)
Helicobacter pylori eradication induces platelet recovery in a subgroup of patients with chronic immune thrombocytopenia (cITP), but the mechanisms involved are still not understood. We aimed to evaluate the effect of H. pylori eradication on platelet response and to identify the
Yifan Hong et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(11), 3211-3216 (2015-03-05)
Glutathione and multidrug resistance protein (MRP) play an important role on the metabolism of a variety of drugs. Bismuth drugs have been used to treat gastrointestinal disorder and Helicobacter pylori infection for decades without exerting acute toxicity. They were found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service