Skip to Content
Merck
All Photos(1)

Key Documents

F15803

Sigma-Aldrich

Formamidine acetate salt

99%

Synonym(s):

Formamidine acetic acid salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN=CHNH2 · CH3COOH
CAS Number:
Molecular Weight:
104.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

mp

158-161 °C (dec.) (lit.)

SMILES string

[H]C(N)=N.CC(O)=O

InChI

1S/C2H4O2.CH4N2/c1-2(3)4;2-1-3/h1H3,(H,3,4);1H,(H3,2,3)

InChI key

XPOLVIIHTDKJRY-UHFFFAOYSA-N

General description

Formamidine acetate salt is an organic compound commonly used as a reagent in the synthesis of heterocyclic compounds.[1]

Application

Formamidine acetate salt is widely used in the preparation of formamidinium lead triiodide (FAPbI3) perovskites for perovskite solar cells.[2][3][4] It is also used in the synthesis of formamidinium lead bromide nanocrystals for use as emitters in display applications.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

He Huang et al.
Nanomaterials (Basel, Switzerland), 10(1) (2020-01-08)
Metal halide perovskites are promising materials for a range of applications. The synthesis of light-emitting perovskite nanorods has become popular recently. Thus far, the facile synthesis of perovskite nanorods remains elusive. In this work, we have developed a facile synthesis
Additive-modulated evolution of HC (NH2) 2PbI3 black polymorph for mesoscopic perovskite solar cells
Wang Z, et al.
Chemistry of Materials, 27(20), 7149-7155 (2015)
Monodisperse formamidinium lead bromide nanocrystals with bright and stable green photoluminescence
Protesescu L, et al.
Journal of the American Chemical Society, 138(43), 14202-14205 (2016)
Nanowire lasers of formamidinium lead halide perovskites and their stabilized alloys with improved stability
Fu Y, et al.
Nano Letters, 16(2), 1000-1008 (2016)
Single crystal formamidinium lead iodide (FAPbI3): Insight into the structural, optical, and electrical properties
Han Q, et al.
Advanced Materials, 28(11), 2253-2258 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service