E33505
Ethyl isonipecotate
98%
Synonym(s):
Ethyl 4-piperidinecarboxylate, Piperidin-4-carboxylic acid ethyl ester
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C8H15NO2
CAS Number:
Molecular Weight:
157.21
Beilstein:
118419
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.459 (lit.)
bp
204 °C (lit.)
density
1.02 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)C1CCNCC1
InChI
1S/C8H15NO2/c1-2-11-8(10)7-3-5-9-6-4-7/h7,9H,2-6H2,1H3
InChI key
RUJPPJYDHHAEEK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Reactant for synthesis of:
Reactant for one-pot reductive amination and Suzuki-Miyaura cross coupling of formyl aryl and heteroaryl MIDA boronates
- SMN protein modulators
- β-aryl and β-amino-substituted aliphatic esters by rhodium catalyzed tandem double bond migration/conjugate addition
- Nitroethylenediamines by nucleophilic ring opening of nitroimidazolidinone
- RhoA inhibitors for cardiovascular disease therapy
- Saccharin derived Mannich bases as antimicrobials and antioxidants
Reactant for one-pot reductive amination and Suzuki-Miyaura cross coupling of formyl aryl and heteroaryl MIDA boronates
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
176.0 °F - closed cup
Flash Point(C)
80 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service